siRNA Details
| Gene Symbol: Smgc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198927.3 | NP_945121.1 | 8 | 924 | ||
| GenBank mRNA | AK017450.1 | - | 8 | 924 | ||
| AK132579.1 | - | 8 | 926 | |||
| AY459348.1 | - | 8 | 925 | |||
| BC049243.1 | - | 3 | 289 | |||
| BC069958.1 | - | 2 | 116 | |||
| BC092073.1 | - | 7 | 745 | |||
| BC138091.1 | - | 8 | 925 | 8861110 | ||
| BC145136.1 | - | 5 | 565 | 9053684 | ||

Smgc
Genomic Map