siRNA Details
| Gene Symbol: Wbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018765.3 | NP_061235.2 | 2 | 351 | ||
| GenBank mRNA | AF071184.1 | - | 2 | 203 | ||
| AK019160.1 | - | 2 | 85 | |||
| AK135215.1 | - | 2 | 351 | |||
| AK159522.1 | - | 2 | 603 | |||
| AK160559.1 | - | 2 | 88 | |||
| AK161496.1 | - | 2 | 45 | |||
| AK162009.1 | - | 2 | 93 | |||
| BC034851.1 | - | 2 | 46 | 1399984 | ||

Wbp4
Genomic Map