siRNA Details
| Gene Symbol: Wbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018765.3 | NP_061235.2 | 3, 4 | 466 | ||
| GenBank mRNA | AF071184.1 | - | 3, 4 | 318 | ||
| AK019160.1 | - | 3, 4 | 200 | |||
| AK135215.1 | - | 3, 4 | 466 | |||
| AK159522.1 | - | 3, 4 | 718 | |||
| AK160559.1 | - | 3, 4 | 203 | |||
| AK161496.1 | - | 3, 4 | 160 | |||
| AK162009.1 | - | 3, 4 | 208 | |||
| BC034851.1 | - | 3, 4 | 161 | 1399984 | ||

Wbp4
Genomic Map