siRNA Details
| Gene Symbol: Adamts20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164785.1 | NP_001158257.1 | 8 | 1403 | ||
| NM_001164786.1 | NP_001158258.1 | 8 | 1403 | |||
| NM_177431.4 | NP_803180.3 | 8 | 1403 | |||
| GenBank mRNA | AJ512753.3 | - | 8 | 1172 | ||
| AK090358.1 | - | 8 | 1369 | |||
| AK145288.1 | - | 8 | 1404 | |||
| AY189815.1 | - | 8 | 1311 | |||
| AY189816.1 | - | 8 | 1311 | |||
| BC121792.1 | - | 8 | 1242 | 40099393 | ||

Adamts20
Genomic Map