siRNA Details
| Gene Symbol: Adamts20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177431.4 | NP_803180.3 | 29 | 4575 | ||
| GenBank mRNA | AJ512753.3 | - | 29 | 4344 | ||
| AY189815.1 | - | 29 | 4483 | |||
| BC121792.1 | - | 29 | 4414 | 40099393 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adamts20| Gene Symbol: Adamts20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177431.4 | NP_803180.3 | 29 | 4575 | ||
| GenBank mRNA | AJ512753.3 | - | 29 | 4344 | ||
| AY189815.1 | - | 29 | 4483 | |||
| BC121792.1 | - | 29 | 4414 | 40099393 | ||