siRNA Details
| Gene Symbol: Adamts20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY189815.1 | - | 34 | 5341 | ||
| BC121792.1 | - | 34 | 5272 | 40099393 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adamts20| Gene Symbol: Adamts20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY189815.1 | - | 34 | 5341 | ||
| BC121792.1 | - | 34 | 5272 | 40099393 | ||