siRNA Details
| Gene Symbol: Senp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144851.5 | NP_659100.1 | 8 | 1232 | ||
| GenBank mRNA | AK028565.1 | - | 8 | 974 | ||
| AK035581.1 | - | 8 | 1068 | |||
| AK035723.1 | - | 8 | 1337 | |||
| AK054521.1 | - | 9 | 1364 | |||
| AK088597.1 | - | 8 | 976 | |||
| AK138013.1 | - | 8 | 1033 | |||
| BC023129.1 | - | 7 | 946 | 3595534 | ||

Senp1
Genomic Map