siRNA Details
| Gene Symbol: Wee1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009516.3 | NP_033542.2 | 1, 2 | 863 | ||
| GenBank mRNA | AK011212.1 | - | 1, 2 | 639 | ||
| AK039572.1 | - | 1, 2 | 179 | |||
| BC006852.1 | - | 1, 2 | 882 | 3156953 | ||
| D30743.1 | - | 1, 2 | 864 | |||

Wee1
Genomic Map