siRNA Details
| Gene Symbol: Spryd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033277.3 | NP_001028449.2 | 7, 8 | 902 | ||
| GenBank mRNA | AK146689.1 | - | 7, 8 | 905 | ||
| BC008150.1 | - | 3, 4 | 413 | |||
| BC048820.1 | - | 6, 7 | 1131 | |||
| CX201967.1 | - | 3, 4 | 401 | |||

Spryd3
Genomic Map