siRNA Details
Gene Symbol: BC048502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177631.3 | NP_808299.1 | 4, 5 | 437 | ||
GenBank mRNA | AK131692.1 | - | 4, 5 | 449 | ||
BC048502.1 | - | 4, 5 | 437 | 6740942 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BC048502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177631.3 | NP_808299.1 | 4, 5 | 437 | ||
GenBank mRNA | AK131692.1 | - | 4, 5 | 449 | ||
BC048502.1 | - | 4, 5 | 437 | 6740942 |