siRNA Details
| Gene Symbol: Wfs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011716.2 | NP_035846.1 | 7, 8 | 993 | ||
| GenBank mRNA | AF084482.1 | - | 7, 8 | 926 | ||
| AJ011971.1 | - | 7, 8 | 920 | |||
| AK145023.1 | - | 7, 8 | 965 | |||
| AK170180.1 | - | 7, 8 | 948 | |||
| BC046988.1 | - | 7, 8 | 929 | 6492038 | ||
| CX562372.1 | - | 5, 6 | 623 | |||

Wfs1
Genomic Map