siRNA Details
| Gene Symbol: Wfs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011716.2 | NP_035846.1 | 8 | 1356 | ||
| GenBank mRNA | AF084482.1 | - | 8 | 1289 | ||
| AJ011971.1 | - | 8 | 1283 | |||
| AK144561.1 | - | 8 | 1080 | |||
| AK145023.1 | - | 8 | 1328 | |||
| AK170180.1 | - | 8 | 1311 | |||
| BC046988.1 | - | 8 | 1292 | 6492038 | ||
| Gene Symbol: Sec14l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK196113.1 | - | 29 | |||

Wfs1
Genomic Map