siRNA Details
| Gene Symbol: Marf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081154.2 | NP_001074623.1 | 25 | 4958 | ||
| GenBank mRNA | AK053159.1 | - | 5 | 676 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Marf1| Gene Symbol: Marf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081154.2 | NP_001074623.1 | 25 | 4958 | ||
| GenBank mRNA | AK053159.1 | - | 5 | 676 | ||