siRNA Details
| Gene Symbol: Pi4ka | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001983.2 | NP_001001983.2 | 50 | 5641 | ||
| GenBank mRNA | AK136942.1 | - | 14 | 1428 | ||
| BC022999.1 | - | 5 | 715 | |||
| BC049252.1 | - | 10 | 979 | |||
| BC055479.1 | - | 4 | 353 | |||
| BC075629.1 | - | 50 | 5641 | 6856662 | ||

Pi4ka
Genomic Map