siRNA Details
| Gene Symbol: Klhl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145479.4 | NP_663454.3 | 6, 7 | 1666 | ||
| GenBank mRNA | AK028197.1 | - | 6, 7 | 2165 | ||
| AK143514.1 | - | 6, 7 | 1611 | |||
| AK214531.1 | - | 1, 2 | 130 | |||
| BC005800.1 | - | 6, 7 | 1644 | 2812589 | ||

Klhl22
Genomic Map