siRNA Details
| Gene Symbol: Klhl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145479.4 | NP_663454.3 | 4 | 750 | ||
| GenBank mRNA | AK028197.1 | - | 4 | 1249 | ||
| AK030669.1 | - | 4 | 695 | |||
| AK086748.1 | - | 4 | 697 | |||
| AK143514.1 | - | 4 | 695 | |||
| BC005800.1 | - | 4 | 728 | 2812589 | ||

Klhl22
Genomic Map