siRNA Details
| Gene Symbol: Fam43a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177632.3 | NP_808300.1 | 1 | 1367 | ||
| GenBank mRNA | AK083036.1 | - | 1 | 1367 | ||
| AK137950.1 | - | 1 | 1368 | |||
| BC099947.1 | - | 1 | 1380 | 30088416 | ||
| BC138882.1 | - | 1 | 981 | 8861904 | ||
| BC138884.1 | - | 1 | 957 | 8861906 | ||

Fam43a
Genomic Map