siRNA Details
Gene Symbol: Poglut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300827.1 | NP_001287756.1 | 11 | 1395 | ||
NM_172380.4 | NP_759012.1 | 11 | 1192 | |||
GenBank mRNA | AK031608.1 | - | 11 | 1131 | ||
AK035948.1 | - | 11 | 1140 | |||
AK036224.1 | - | 11 | 1122 | |||
AK041321.1 | - | 11 | 1130 | |||
AK045278.1 | - | 11 | 1340 | |||
AK079886.1 | - | 11 | 1162 | |||
AK167641.1 | - | 11 | 1362 | |||
AK216969.1 | - | 3 | 246 | |||
BC026809.1 | - | 11 | 1143 | 4205469 |
