siRNA Details
Gene Symbol: Poglut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300827.1 | NP_001287756.1 | 7, 8 | 1019 | ||
NM_172380.4 | NP_759012.1 | 7, 8 | 816 | |||
GenBank mRNA | AK031608.1 | - | 7, 8 | 755 | ||
AK035948.1 | - | 7, 8 | 764 | |||
AK036224.1 | - | 7, 8 | 746 | |||
AK041321.1 | - | 7, 8 | 754 | |||
AK045278.1 | - | 7, 8 | 964 | |||
AK167641.1 | - | 7, 8 | 986 | |||
BC026809.1 | - | 7, 8 | 767 | 4205469 |
