siRNA Details
| Gene Symbol: Dzip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110017.1 | NP_001103487.1 | 22 | 2874 | ||
| NM_027341.2 | NP_081617.1 | 21 | 2256 | |||
| GenBank mRNA | BC117953.1 | - | 21 | 2079 | 40098141 | |
| BC117954.1 | - | 21 | 2085 | 40098143 | ||
| EU233997.1 | - | 14 | 2076 | |||

Dzip3
Genomic Map