siRNA Details
| Gene Symbol: Dzip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110017.1 | NP_001103487.1 | 19 | 2463 | ||
| NM_027341.2 | NP_081617.1 | 18 | 1845 | |||
| GenBank mRNA | AK039172.1 | - | 19 | 2441 | ||
| BC117953.1 | - | 18 | 1668 | 40098141 | ||
| BC117954.1 | - | 18 | 1674 | 40098143 | ||
| EU233997.1 | - | 11 | 1665 | |||

Dzip3
Genomic Map