siRNA Details
| Gene Symbol: Impg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_174876.3 | NP_777365.2 | 18 | 3775 | ||
| GenBank mRNA | AK047356.1 | - | 4 | 666 | ||
| AY174061.1 | - | 17 | 3448 | |||
| BC026456.1 | - | 4 | 661 | |||
| BC048863.1 | - | 18 | 3744 | 5368568 | ||

Impg2
Genomic Map