siRNA Details
Gene Symbol: Crybg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174848.3 | NP_777273.2 | 2, 3 | 1687 | ||
GenBank mRNA | BC043118.1 | - | 2, 3 | 1687 | 6408859 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Crybg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174848.3 | NP_777273.2 | 2, 3 | 1687 | ||
GenBank mRNA | BC043118.1 | - | 2, 3 | 1687 | 6408859 |