siRNA Details
| Gene Symbol: Wt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144783.2 | NP_659032.3 | 10 | 1780 | ||
| GenBank mRNA | DQ537939.1 | - | 10 | 1117 | ||
| M55512.1 | - | 10 | 1785 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wt1| Gene Symbol: Wt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144783.2 | NP_659032.3 | 10 | 1780 | ||
| GenBank mRNA | DQ537939.1 | - | 10 | 1117 | ||
| M55512.1 | - | 10 | 1785 | |||