siRNA Details
| Gene Symbol: Wt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144783.2 | NP_659032.3 | 6, 7 | 1370 | ||
| GenBank mRNA | BG087452.2 | - | 2, 3 | 169 | ||
| DQ537939.1 | - | 6, 7 | 707 | |||
| M55512.1 | - | 6, 7 | 1375 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wt1| Gene Symbol: Wt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144783.2 | NP_659032.3 | 6, 7 | 1370 | ||
| GenBank mRNA | BG087452.2 | - | 2, 3 | 169 | ||
| DQ537939.1 | - | 6, 7 | 707 | |||
| M55512.1 | - | 6, 7 | 1375 | |||