siRNA Details
| Gene Symbol: Scaf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178923.3 | NP_849254.2 | 6 | 967 | ||
| GenBank mRNA | AK045266.1 | - | 6 | 964 | ||
| AK048239.1 | - | 5 | 1618 | |||
| AK173120.1 | - | 6 | 1045 | |||
| AK216612.1 | - | 3 | 313 | |||
| BC023734.1 | - | 3 | 368 | |||
| BC041782.1 | - | 3 | 339 | |||
| BC053096.1 | - | 6 | 862 | 6401143 | ||
| BC057592.1 | - | 6 | 967 | 6417362 | ||

Scaf4
Genomic Map