siRNA Details
| Gene Symbol: Slc37a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242427.1 | NP_001229356.1 | 4 | 1039 | ||
| NM_153062.2 | NP_694702.1 | 4 | 619 | |||
| GenBank mRNA | AK033656.1 | - | 4 | 508 | ||
| AK143636.1 | - | 4 | 1045 | |||
| AK153706.1 | - | 4 | 1042 | |||
| AK163990.1 | - | 3 | 221 | |||
| BC027294.1 | - | 4 | 619 | 3985469 | ||

Slc37a1
Genomic Map