siRNA Details
| Gene Symbol: Vps52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172620.3 | NP_766208.2 | 6 | 676 | ||
| GenBank mRNA | AK042520.1 | - | 6 | 630 | ||
| AK051732.1 | - | 6 | 747 | |||
| AK052523.1 | - | 6 | 677 | |||
| AK210671.1 | - | 2 | 164 | |||
| AK214507.1 | - | 2 | 168 | |||
| AX775735.1 | - | 6 | 488 | |||
| KJ650214.1 | - | 6 | 465 | |||

Vps52
Genomic Map