siRNA Details
| Gene Symbol: Ubr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177374.1 | NP_001170845.1 | 25, 26 | 3079 | ||
| NM_146078.3 | NP_666190.2 | 25, 26 | 3079 | |||
| GenBank mRNA | AK083320.1 | - | 25, 26 | 3218 | ||
| AK122254.2 | - | 23, 24 | 3908 | |||
| AK143202.1 | - | 25, 26 | 3079 | |||
| AK147445.1 | - | 25, 26 | 3068 | |||
| AY061885.1 | - | 25, 26 | 2766 | |||
| AY280958.1 | - | 25, 26 | 2794 | |||
| BC031403.1 | - | 10, 11 | 873 | 4935220 | ||
| BC044903.1 | - | 11, 12 | 1014 | |||
| BC075642.1 | - | 26, 27 | 2893 | 30544183 | ||

Ubr2
Genomic Map