siRNA Details
Gene Symbol: Map4k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290345.1 | NP_001277274.1 | 7 | 574 | ||
GenBank mRNA | AK014711.1 | - | 7 | 563 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Map4k3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290345.1 | NP_001277274.1 | 7 | 574 | ||
GenBank mRNA | AK014711.1 | - | 7 | 563 |