siRNA Details
Gene Symbol: Svil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153153.3 | NP_694793.1 | 3 | 344 | ||
GenBank mRNA | AF317422.1 | - | 3 | 359 | ||
AF317423.1 | - | 3 | 359 | |||
AK021019.1 | - | 6 | 833 | |||
AK041925.1 | - | 6 | 819 | |||
BC115653.1 | - | 6 | 614 | |||
BC115654.1 | - | 6 | 614 | |||
BC128496.1 | - | 6 | 614 | |||
BC128499.1 | - | 6 | 614 | |||
BC152345.1 | - | 6 | 614 | |||
BC152346.1 | - | 6 | 614 | |||
HQ259291.1 | - | 3 | 359 |
