siRNA Details
| Gene Symbol: Gjd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153086.5 | NP_694726.2 | 2 | 1093 | ||
| GenBank mRNA | AJ414562.1 | - | 2 | 902 | ||
| AK034663.1 | - | 2 | 1094 | |||
| AK053016.1 | - | 2 | 1045 | |||
| BC137984.1 | - | 2 | 1070 | 8861003 | ||

Gjd4
Genomic Map