siRNA Details
| Gene Symbol: Gjd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153086.5 | NP_694726.2 | 2 | 468 | ||
| GenBank mRNA | AJ414562.1 | - | 2 | 277 | ||
| AK034663.1 | - | 2 | 469 | |||
| AK053016.1 | - | 2 | 420 | |||
| BC137984.1 | - | 2 | 445 | 8861003 | ||

Gjd4
Genomic Map