siRNA Details
Gene Symbol: Mib1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144860.2 | NP_659109.2 | 15 | 2755 | ||
GenBank mRNA | AK053035.1 | - | 8 | 978 | ||
AK080847.1 | - | 9 | 1031 | |||
AY147848.1 | - | 15 | 2254 | |||
AY149907.1 | - | 15 | 2075 | |||
AY245539.1 | - | 15 | 2755 | |||
AY974091.1 | - | 15 | 2048 | |||
BC011287.1 | - | 4 | 352 | |||
BC083072.1 | - | 5 | 433 | |||
BC131662.1 | - | 16 | 2221 | 40110696 |
