siRNA Details
| Gene Symbol: Zfp521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145492.4 | NP_663467.1 | 6, 7 | 4020 | ||
| GenBank mRNA | AK052655.1 | - | 6, 7 | 3924 | ||
| AK077697.1 | - | 3, 4 | 842 | |||
| AK081096.1 | - | 4, 5 | 1474 | |||
| AK131130.1 | - | 5, 6 | 4105 | |||
| AY147406.1 | - | 6, 7 | 3923 | |||
| BC021376.1 | - | 6, 7 | 3836 | 5038671 | ||
| BC023458.1 | - | 3, 4 | 556 | |||

Zfp521
Genomic Map