siRNA Details
| Gene Symbol: Zfp521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145492.4 | NP_663467.1 | 4 | 3741 | ||
| GenBank mRNA | AK052655.1 | - | 4 | 3645 | ||
| AK077697.1 | - | 1 | 563 | |||
| AK081096.1 | - | 1 | 1198 | |||
| AK131130.1 | - | 3 | 3826 | |||
| AY147406.1 | - | 4 | 3644 | |||
| BC021376.1 | - | 4 | 3557 | 5038671 | ||
| BC023458.1 | - | 1 | 277 | |||

Zfp521
Genomic Map