siRNA Details
| Gene Symbol: Ammecr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242430.1 | NP_001229359.1 | 3, 4 | 1135 | ||
| NM_153515.4 | NP_705735.1 | 3, 4 | 934 | |||
| GenBank mRNA | AK030009.1 | - | 3, 4 | 1142 | ||
| AK054131.1 | - | 3, 4 | 933 | |||
| AK133018.1 | - | 3, 4 | 1136 | |||
| AK208687.1 | - | 1, 2 | 144 | |||
| BC012310.1 | - | 2, 3 | 539 | |||
| BC034661.1 | - | 3, 4 | 675 | 5137474 | ||
| BC056948.1 | - | 3, 4 | 878 | 6827318 | ||

Ammecr1l
Genomic Map