siRNA Details
| Gene Symbol: Lvrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029008.1 | NP_083284.1 | 10 | 1877 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lvrn| Gene Symbol: Lvrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029008.1 | NP_083284.1 | 10 | 1877 | ||