siRNA Details
| Gene Symbol: A730017C20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167925.2 | NP_001161397.2 | 4 | 616 | ||
| NM_173759.5 | NP_776120.3 | 3 | 565 | |||
| GenBank mRNA | AK080452.1 | - | 5 | 569 | ||
| BC075669.1 | - | 5 | 525 | 30550997 | ||

A730017C20Rik
Genomic Map