siRNA Details
| Gene Symbol: A730017C20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167925.2 | NP_001161397.2 | 3 | 420 | ||
| NM_173759.5 | NP_776120.3 | 2 | 369 | |||
| GenBank mRNA | AK048493.1 | - | 3 | 1398 | ||
| AK080452.1 | - | 4 | 373 | |||
| AK086069.1 | - | 1 | 1481 | |||
| BC075669.1 | - | 4 | 329 | 30550997 | ||
| BY125990.1 | - | 2 | 224 | |||

A730017C20Rik
Genomic Map