siRNA Details
| Gene Symbol: Pde6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146086.2 | NP_666198.1 | 1 | 123 | ||
| GenBank mRNA | BC031925.1 | - | 1 | 123 | 4501123 | |
| BC044892.1 | - | 1 | 123 | 4527430 | ||
| BC158126.1 | - | 1 | 307 | 9007394 | ||
| X60664.1 | - | 1 | 131 | |||

Pde6a
Genomic Map