siRNA Details
| Gene Symbol: Piezo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039485.4 | NP_001034574.4 | 45 | 7958 | ||
| GenBank mRNA | AK031235.1 | - | 5 | 896 | ||
| AK031422.1 | - | 4 | 617 | |||
| BC026614.1 | - | 2 | 346 | |||
| BC075677.1 | - | 6 | 1049 | |||
| BC147606.1 | - | 4 | 511 | 9056596 | ||
| BC172109.1 | - | 4 | 511 | |||
| HQ215521.1 | - | 45 | 7038 | |||

Piezo2
Genomic Map