siRNA Details
| Gene Symbol: Mapk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172632.2 | NP_766220.2 | 4, 5 | 1799 | ||
| GenBank mRNA | AK079654.1 | - | 4, 5 | 1777 | ||
| BC058942.1 | - | 3, 4 | 1202 | 6821544 | ||
| BC062911.1 | - | 3, 4 | 1202 | 6821544 | ||
| BC094564.1 | - | 3, 4 | 2013 | |||

Mapk4
Genomic Map