siRNA Details
| Gene Symbol: Zadh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146090.5 | NP_666202.2 | 2 | 380 | ||
| GenBank mRNA | AK047559.1 | - | 2 | 385 | ||
| AK081988.1 | - | 2 | 381 | |||
| AK083082.1 | - | 2 | 424 | |||
| BC116427.1 | - | 2 | 381 | 40104327 | ||
| BC116428.1 | - | 2 | 381 | 40104329 | ||

Zadh2
Genomic Map