siRNA Details
| Gene Symbol: Catsper1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139301.2 | NP_647462.1 | 1 | 705 | ||
| GenBank mRNA | AF407332.1 | - | 1 | 705 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Catsper1| Gene Symbol: Catsper1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139301.2 | NP_647462.1 | 1 | 705 | ||
| GenBank mRNA | AF407332.1 | - | 1 | 705 | ||