siRNA Details
| Gene Symbol: Kdm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001984.2 | NP_001001984.2 | 3 | 960 | ||
| GenBank mRNA | AK037157.1 | - | 4 | 857 | ||
| AK147028.1 | - | 3 | 952 | |||
| AK155866.1 | - | 3 | 965 | |||
| BC076576.1 | - | 3 | 526 | 6840591 | ||

Kdm2a
Genomic Map