siRNA Details
| Gene Symbol: Atrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009530.2 | NP_033556.2 | 29, 30 | 6716 | ||
| GenBank mRNA | AF026032.1 | - | 30, 31 | 6683 | ||
| AK134024.1 | - | 21, 22 | 2841 | |||
| AK161784.1 | - | 18, 19 | 2462 | |||

Atrx
Genomic Map