siRNA Details
| Gene Symbol: Atrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009530.2 | NP_033556.2 | 15, 16 | 4761 | ||
| GenBank mRNA | AF026032.1 | - | 16, 17 | 4728 | ||
| AK134024.1 | - | 7, 8 | 886 | |||
| AK161784.1 | - | 4, 5 | 507 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atrx| Gene Symbol: Atrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009530.2 | NP_033556.2 | 15, 16 | 4761 | ||
| GenBank mRNA | AF026032.1 | - | 16, 17 | 4728 | ||
| AK134024.1 | - | 7, 8 | 886 | |||
| AK161784.1 | - | 4, 5 | 507 | |||