siRNA Details
| Gene Symbol: Atrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009530.2 | NP_033556.2 | 18, 19 | 5174 | ||
| GenBank mRNA | AF026032.1 | - | 19, 20 | 5141 | ||
| AK134024.1 | - | 10, 11 | 1299 | |||
| AK161784.1 | - | 7, 8 | 920 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atrx| Gene Symbol: Atrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009530.2 | NP_033556.2 | 18, 19 | 5174 | ||
| GenBank mRNA | AF026032.1 | - | 19, 20 | 5141 | ||
| AK134024.1 | - | 10, 11 | 1299 | |||
| AK161784.1 | - | 7, 8 | 920 | |||