siRNA Details
| Gene Symbol: Tkfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145496.1 | NP_663471.1 | 8 | 825 | ||
| GenBank mRNA | AK138956.1 | - | 865 | |||
| AK171418.1 | - | 9 | 1016 | |||
| BC021917.1 | - | 8 | 825 | 4481611 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tkfc| Gene Symbol: Tkfc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145496.1 | NP_663471.1 | 8 | 825 | ||
| GenBank mRNA | AK138956.1 | - | 865 | |||
| AK171418.1 | - | 9 | 1016 | |||
| BC021917.1 | - | 8 | 825 | 4481611 | ||